Anti-RELA Rabbit Polyclonal Antibody
Catalog # ORIGTA319207
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Transcription factor p65
- Antigen symbol:RELA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:near C-terminus
- Cross adsorption:No
- Form:Liquid
- Storage buffer:None
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:NFkB p65 (Rel A) peptide corresponding to a region near the C-terminus of the human protein conjugated to Keyhole Limpet Hemocyanin (KLH).
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:2000-1:5000; Immunochemistry: 1:400 Gel ShImmunofluorescencet: 05 ?l-1.0 ?l
Type: Primary
Antigen: RELA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat