Anti-HSP90AA1 Rabbit Polyclonal Antibody
Catalog # ORIGTA319144
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:HSP90AA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Drosophila,Monkey,Chicken,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:289 to 300
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 289-300 of human Hsp90 protein.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: ELISA: 1:5000-1:25000; Western Blot: 1:500-1:2500
Type: Primary
Antigen: HSP90AA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Drosophila, Human, Monkey, Mouse, Rat