Anti-PKM2 Mouse Monoclonal Antibody [clone: 11G10]
Catalog # ORIGTA190269
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:PKM2
- Clonality:Monoclonal
- Clone:11G10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Sequence:LRRLAPITSDPTEATAVGAV
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Synthetic peptide: LRRLAPITSDPTEATAVGAV, corresponding to the region encoded by PKM gene M2 isoform.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: PKM2
Clonality: Monoclonal
Clone: 11G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human