Anti-SHBG Mouse Monoclonal Antibody [clone: 1E7]
Catalog # ORIGCF507137
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Sex Hormone Binding Globulin
- Antigen symbol:SHBG
- Clonality:Monoclonal
- Clone:1E7
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:38.4 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human SHBG(NP_001139751) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:4000
Type: Primary
Antigen: SHBG
Clonality: Monoclonal
Clone: 1E7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human