Order Entry
Export
ContactUsLinkComponent
Anti-ERG Mouse Monoclonal Antibody [clone: 8A9]
Anti-ERG Mouse Monoclonal Antibody [clone: 8A9]
Catalog # ORIGCF506201
Supplier:  OriGene
Anti-ERG Mouse Monoclonal Antibody [clone: 8A9]
Catalog # ORIGCF506201
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    ERG
  • Clonality:
    Monoclonal
  • Clone:
    8A9
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    Full length
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    53.7 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Full lengthu human recombinant protein of human ERG(NP_891548) produced in HEK293T cell.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:50-1:100

Type: Primary
Antigen: ERG
Clonality: Monoclonal
Clone: 8A9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human