Anti-OTC Mouse Monoclonal Antibody [clone: 4G8]
Catalog # ORIGCF802596
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:OTC
- Clonality:Monoclonal
- Clone:4G8
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:33 to 354
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:36.1 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 33-354 of human OTC (NP_000522)produced in SF9 cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: OTC
Clonality: Monoclonal
Clone: 4G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human