Anti-KI67 Mouse Monoclonal Antibody [clone: 9H1]
ORIGCF802431
: OriGene
- Antibody type:Primary
- Antigen symbol:KI67
- Clonality:Monoclonal
- Clone:9H1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:1160 to 1493
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 1160-1493 of human MKI67 (NP_002408) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Immunochemistry: 1:100
Type: Primary
Antigen: KI67
Clonality: Monoclonal
Clone: 9H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human