Anti-CD5 Mouse Monoclonal Antibody [clone: 10C8]
Catalog # ORIGCF802425
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:CD5
- Clonality:Monoclonal
- Clone:10C8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:54.4 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human CD5 (NP_055022) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Flow Cytometry: 1:50
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 10C8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human