Anti-MTIF3 Mouse Monoclonal Antibody [clone: 4B6]
Catalog # ORIGCF800436
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:MTIF3
- Clonality:Monoclonal
- Clone:4B6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Monkey,Human,Dog
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:56 to 278
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:31.5 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 56-278 of human MTIF3 (NP_690876) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: MTIF3
Clonality: Monoclonal
Clone: 4B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey