Anti-CHAF1B Mouse Monoclonal Antibody [clone: 2D5]
ORIGCF506841
: OriGene
- Antibody type:Primary
- Antigen symbol:CHAF1B
- Clonality:Monoclonal
- Clone:2D5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:61.3 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human CHAF1B(NP_005432) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100
Type: Primary
Antigen: CHAF1B
Clonality: Monoclonal
Clone: 2D5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human