Anti-A4GNT Mouse Monoclonal Antibody [clone: 1F7]
Catalog # ORIGCF800401
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:alpha-1,4-N-Acetylglucosaminyltransferase
- Antigen symbol:A4GNT
- Clonality:Monoclonal
- Clone:1F7
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Monkey,Human,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:121 to 340
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:39.3 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 121-340 of human A4GNT (NP_057245) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: A4GNT
Clonality: Monoclonal
Clone: 1F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat