Anti-NBN Mouse Monoclonal Antibody [clone: 3G3]
ORIGCF801423
: OriGene
- Antibody type:Primary
- Antigen symbol:NBN
- Clonality:Monoclonal
- Clone:3G3
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:183 to 460
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:84.8 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 183-460 of human NBN (NP_002476)produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: NBN
Clonality: Monoclonal
Clone: 3G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human