Anti-MRPS27 Mouse Monoclonal Antibody [clone: 3B9]
ORIGCF800199
: OriGene
- Antibody type:Primary
- Antigen symbol:MRPS27
- Clonality:Monoclonal
- Clone:3B9
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:151 to 414
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:47.4 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 151-414 of human MRPS27 (NP_055899) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: MRPS27
Clonality: Monoclonal
Clone: 3B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human