Anti-ALK Mouse Monoclonal Antibody [clone: 6E11]
Catalog # ORIGCF801232
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:anaplastic lymphoma receptor tyrosine kinase
- Antigen symbol:ALK
- Clonality:Monoclonal
- Clone:6E11
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:1300 to 1620
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:176.3 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 1300-1620 of human ALK (NP_004295) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: ALK
Clonality: Monoclonal
Clone: 6E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human