Order Entry
Export
ContactUsLinkComponent
 
Anti-SENP2 Mouse Monoclonal Antibody [clone: 3D3]
  ORIGCF800123
 :  OriGene
Anti-SENP2 Mouse Monoclonal Antibody [clone: 3D3]
  ORIGCF800123
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen name:
    Sentrin-specific protease 2
  • Antigen symbol:
    SENP2
  • Clonality:
    Monoclonal
  • Clone:
    3D3
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Monkey,
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    139 to 523
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Human recombinant protein fragment corresponding to amino acids 139-523 of human SENP2 (NP_067640) produced in E.coli.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SENP2
Clonality: Monoclonal
Clone: 3D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey