Anti-USP44 Mouse Monoclonal Antibody [clone: 5B8]
Catalog # ORIGCF801915
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin specific peptidase 44
- Antigen symbol:USP44
- Clonality:Monoclonal
- Clone:5B8
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:420 to 712
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:81 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 420-712 of human USP44 (NP_115523) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: USP44
Clonality: Monoclonal
Clone: 5B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human