Anti-DPPA4 Mouse Monoclonal Antibody [clone: 1E12]
Catalog # ORIGCF800064
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Developmental pluripotency associated 4
- Antigen symbol:DPPA4
- Clonality:Monoclonal
- Clone:1E12
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:9 to 254
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:33.4 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 9-254 of human DPPA4 (NP_060659) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: DPPA4
Clonality: Monoclonal
Clone: 1E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human