Anti-ARL3 Mouse Monoclonal Antibody [clone: 3E1]
ORIGCF803089
: OriGene
- Antibody type:Primary
- Antigen name:ADP-ribosylation factor-like 3
- Antigen symbol:ARL3
- Clonality:Monoclonal
- Clone:3E1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:2 to 182
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:20.3 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 2-182 of human ARL3 (NP_004302) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: ARL3
Clonality: Monoclonal
Clone: 3E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human