Anti-TRIM9 Mouse Monoclonal Antibody [clone: 2H3]
ORIGCF800043
: OriGene
- Antibody type:Primary
- Antigen symbol:TRIM9
- Clonality:Monoclonal
- Clone:2H3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:284 to 669
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:61.2 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 284-669 of human TRIM9 (NP_443210) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: TRIM9
Clonality: Monoclonal
Clone: 2H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human