Anti-UQCRC1 Mouse Monoclonal Antibody [clone: 1G2]
Catalog # ORIGCF800033
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Ubiquinol-cytochrome c reductase core protein 1
- Antigen symbol:UQCRC1
- Clonality:Monoclonal
- Clone:1G2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:46 to 305
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:52.5 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 46-305 of human UQCRC1 (NP_003356) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-1000; Immunofluorescence: 1:100
Type: Primary
Antigen: UQCRC1
Clonality: Monoclonal
Clone: 1G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human