Order Entry
Export
ContactUsLinkComponent
Anti-ACSM5 Mouse Monoclonal Antibody [clone: 1G1]
Anti-ACSM5 Mouse Monoclonal Antibody [clone: 1G1]
  ORIGCF504441
 :  OriGene
Anti-ACSM5 Mouse Monoclonal Antibody [clone: 1G1]
  ORIGCF504441
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen name:
    acyl-CoA synthetase medium-chain family member 5
  • Antigen symbol:
    ACSM5
  • Clonality:
    Monoclonal
  • Clone:
    1G1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    Full length
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    64.6 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Full length human recombinant protein of human ACSM5(NP_060358) produced in HEK293T cell.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: ACSM5
Clonality: Monoclonal
Clone: 1G1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human