Anti-SERPINA5 Mouse Monoclonal Antibody [clone: 1A12]
ORIGCF504357
: OriGene
- Antibody type:Primary
- Antigen name:Serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5
- Antigen symbol:SERPINA5
- Clonality:Monoclonal
- Clone:1A12
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Dog,Mouse
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:45.5 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human SERPINA5(NP_000615) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1:500-2000
Type: Primary
Antigen: SERPINA5
Clonality: Monoclonal
Clone: 1A12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse