Anti-XPNPEP3 Mouse Monoclonal Antibody [clone: 4B8]
ORIGCF503339
: OriGene
- Antibody type:Primary
- Antigen symbol:XPNPEP3
- Clonality:Monoclonal
- Clone:4B8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Monkey,Human,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:56.9 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human XPNPEP3(NP_071381) produced in HEK293 cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: XPNPEP3
Clonality: Monoclonal
Clone: 4B8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat