Anti-LZIC Mouse Monoclonal Antibody [clone: 3A7]
Catalog # ORIGCF505823
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:LZIC
- Clonality:Monoclonal
- Clone:3A7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:1 to 190
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:21.3 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 1-190 of human LZIC (NP_115744) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:400-1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: LZIC
Clonality: Monoclonal
Clone: 3A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human