Anti-ERBB2 Mouse Monoclonal Antibody [clone: C4]
Catalog # ORIGCF505819
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:ERBB2
- Clonality:Monoclonal
- Clone:C4
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:676 to 1255
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:137.7 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 676-1255 of human ERBB2(NP_004439) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:200-1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human