Order Entry
Export
ContactUsLinkComponent
Anti-ERBB2 Mouse Monoclonal Antibody [clone: 6H10]
Anti-ERBB2 Mouse Monoclonal Antibody [clone: 6H10]
Catalog # ORIGCF505792
Supplier:  OriGene
Anti-ERBB2 Mouse Monoclonal Antibody [clone: 6H10]
Catalog # ORIGCF505792
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    ERBB2
  • Clonality:
    Monoclonal
  • Clone:
    6H10
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    676 to 1255
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    137.7 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Human recombinant protein fragment corresponding to amino acids 676-1255 of human ERBB2(NP_004439) produced in HEK293T cell.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: 6H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human