Anti-SPINT1 Mouse Monoclonal Antibody [clone: 4E3]
Catalog # ORIGCF504562
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:SPINT1
- Clonality:Monoclonal
- Clone:4E3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human SPINT1(NP_857593) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: SPINT1
Clonality: Monoclonal
Clone: 4E3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human