Anti-ALDH3A2 Mouse Monoclonal Antibody [clone: 2A7]
ORIGCF503256
: OriGene
- Antibody type:Primary
- Antigen name:aldehyde dehydrogenase 3 family, member A2
- Antigen symbol:ALDH3A2
- Clonality:Monoclonal
- Clone:2A7
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:57.5 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human ALDH3A2(NP_001026976) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: ALDH3A2
Clonality: Monoclonal
Clone: 2A7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human