Order Entry
Export
ContactUsLinkComponent
Anti-SIGLEC9 Mouse Monoclonal Antibody [clone: 1D9]
Anti-SIGLEC9 Mouse Monoclonal Antibody [clone: 1D9]
Catalog # ORIGCF500379
Supplier:  OriGene
Anti-SIGLEC9 Mouse Monoclonal Antibody [clone: 1D9]
Catalog # ORIGCF500379
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sialic acid binding Ig-like lectin 9
  • Antigen symbol:
    SIGLEC9
  • Clonality:
    Monoclonal
  • Clone:
    1D9
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Monkey,
    Human,
    Dog
  • Western blot:
    Yes
  • Epitope:
    full length
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    50.1 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Full length human recombinant protein of human SIGLEC9 (NP_055256) produced in HEK293T cell.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1:20000-40000; Immunochemistry: 1:50

Type: Primary
Antigen: SIGLEC9
Clonality: Monoclonal
Clone: 1D9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey