Anti-PPM1G Mouse Monoclonal Antibody [clone: 1C12]
ORIGCF502232
: OriGene
- Antibody type:Primary
- Antigen symbol:PPM1G
- Clonality:Monoclonal
- Clone:1C12
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human,Dog
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:59.1 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human PPM1G (NP_817092) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100
Type: Primary
Antigen: PPM1G
Clonality: Monoclonal
Clone: 1C12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human