Order Entry
Export
ContactUsLinkComponent
 
Anti-JNK1 Mouse Monoclonal Antibody [clone: 4E3]
Catalog # ORIGCF500081
Supplier:  OriGene
Anti-JNK1 Mouse Monoclonal Antibody [clone: 4E3]
Catalog # ORIGCF500081
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    JNK1
  • Clonality:
    Monoclonal
  • Clone:
    4E3
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    1 to 384
  • Storage buffer:
    PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    44.0 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Human recombinant protein fragment corresponding to amino acids 1-384 of human JNK1 (NP_002741) produced in E.coli.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: JNK1
Clonality: Monoclonal
Clone: 4E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human