Anti-ACAT2 Mouse Monoclonal Antibody [clone: 1B7]
Catalog # ORIGCF501238
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Acetyl-CoA acetyltransferase 2
- Antigen symbol:ACAT2
- Clonality:Monoclonal
- Clone:1B7
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Monkey,Human,Dog,Rat
- Western blot:Yes
- Epitope:full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:41.2 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human ACAT2 (NP_0058826) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: ACAT2
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Rat