Anti-NXNL2 Mouse Monoclonal Antibody [clone: 1C3]
Catalog # ORIGCF501215
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:NXNL2
- Clonality:Monoclonal
- Clone:1C3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human,Dog
- Western blot:Yes
- Epitope:full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:14.5 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human NXNL2 (NP_660326) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100
Type: Primary
Antigen: NXNL2
Clonality: Monoclonal
Clone: 1C3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human