Anti-ANXA3 Mouse Monoclonal Antibody [clone: 1A5]
Catalog # ORIGCF502123
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Annexin A3
- Antigen symbol:ANXA3
- Clonality:Monoclonal
- Clone:1A5
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:Lipocortin III
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:36.2 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human ANXA3 (NP_005130) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: ANXA3
Clonality: Monoclonal
Clone: 1A5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat