Order Entry
Export
ContactUsLinkComponent
Anti-SDR9C7 Mouse Monoclonal Antibody [clone: 11H10]
Anti-SDR9C7 Mouse Monoclonal Antibody [clone: 11H10]
Catalog # ORIGCF501734
Supplier:  OriGene
Anti-SDR9C7 Mouse Monoclonal Antibody [clone: 11H10]
Catalog # ORIGCF501734
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Short Chain Dehydrogenase/Reductase Family 9C Member 7
  • Antigen symbol:
    SDR9C7
  • Clonality:
    Monoclonal
  • Clone:
    11H10
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    Full length
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    35.1 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Full length human recombinant protein of human SDR9C7 (NP_683695) produced in E.coli.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: SDR9C7
Clonality: Monoclonal
Clone: 11H10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human