Anti-SDR9C7 Mouse Monoclonal Antibody [clone: 4B5]
ORIGCF501723
: OriGene
- Antibody type:Primary
- Antigen name:Short Chain Dehydrogenase/Reductase Family 9C Member 7
- Antigen symbol:SDR9C7
- Clonality:Monoclonal
- Clone:4B5
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:35.1 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human SDR9C7 (NP_683695) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: SDR9C7
Clonality: Monoclonal
Clone: 4B5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human