Anti-CHCHD5 Mouse Monoclonal Antibody [clone: 9E11]
Catalog # ORIGCF502668
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Coiled-coil-helix-coiled-coil-helix domain containing 5
- Antigen symbol:CHCHD5
- Clonality:Monoclonal
- Clone:9E11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:12.2 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human CHCHD5 (NP_115685) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: CHCHD5
Clonality: Monoclonal
Clone: 9E11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human