Anti-XRCC1 Mouse Monoclonal Antibody [clone: 2D8]
ORIGCF500886
: OriGene
- Antibody type:Primary
- Antigen symbol:XRCC1
- Clonality:Monoclonal
- Clone:2D8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:69.3 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human XRCC1 (NP_006288) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Flow Cytometry: 1:100
Type: Primary
Antigen: XRCC1
Clonality: Monoclonal
Clone: 2D8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human