Anti-CSF3 Rabbit Polyclonal Antibody
Catalog # ORIGTA319031
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:CSF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:100 µg (0,2 mg/ml) affinity purified rabbit anti-human G-CSF polyclonal antibody in phosphate (PBS, pH 7,2) containing 30% glycerol, 0,5 % BSA and 0,01% thimerosal.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Synthetic peptide surrounding amino acid 195 of human G-CSF
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1-4 ?g/ml
Type: Primary
Antigen: CSF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human