Anti-ATG9B Rabbit Polyclonal Antibody
Catalog # ORIGTA319026
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:autophagy related 9B
- Antigen symbol:ATG9B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Form:Liquid
- Storage buffer:100 µg (1 mg/ml) in 1X PBS containing 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:ATG9B antibody was raised against a 17 amino acid peptide near the carboxy terminus of human ATG9B.
- Purification:purified by affinity chromatography via peptide column
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunochemistry: 10?g/ml;
Type: Primary
Antigen: ATG9B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat