Order Entry
Export
ContactUsLinkComponent
Anti-Slamf8 Rabbit Polyclonal Antibody
Anti-Slamf8 Rabbit Polyclonal Antibody
Catalog # ORIGTA318999
Supplier:  OriGene
Anti-Slamf8 Rabbit Polyclonal Antibody
Catalog # ORIGTA318999
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Signaling lymphocytic activation molecule family member 8
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    100 µg (0,5 mg/ml) affinity purified rabbit polyclonal antibody in phosphate-buffered saline (PBS) containing 30% glycerol, 0,5% BSA, and 0,01% thimerosal.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Synthetic peptide surrounding amino acid 40 of rat BLAME
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 0.5-4 ?g/ml

Type: Primary
Antigen: Slamf8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat