Order Entry
Export
ContactUsLinkComponent
Anti-TLR5 Rabbit Polyclonal Antibody
Anti-TLR5 Rabbit Polyclonal Antibody
  BSBTPB9451
undefined
Anti-TLR5 Rabbit Polyclonal Antibody
  BSBTPB9451

 

  • Antibody type:
    Primary
  • Antigen name:
    Toll-like Receptor 5
  • Antigen symbol:
    TLR5
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Gene ID:
    7100
  • Antigen synonyms:
    SLEB1|TIL3|MELIOS|SLE1
  • UniProtKB:
    O60602
  • Storage temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human TLR5 (801-828aa MKHQSIRGFVQKQQYLRWPEDFQDVGWF), different from the related mouse sequence by five amino acids.
  • Purification:
    Immunogen affinity purified.
  • Size:
    100 µg/vial
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Toll-like receptor 5(TLR5) detection. Tested with WB, IHC-P in Human.

TLR5 encodes a member of the toll-like receptor (TLR) family, which plays a fundamental role in pathogen recognition and activation of innate immune responses. These receptors recognize distinct pathogen-associated molecular patterns that are expressed on infectious agents. The protein encoded by this gene recognizes bacterial flagellin, the principal component of bacterial flagella and a virulence factor. The activation of this receptor mobilizes the nuclear factor NF-kappaB, which in turn activates a host of inflammatory-related target genes. Mutations in this gene have been associated with both resistance and susceptibility to systemic lupus erythematosus, and susceptibility to Legionnaire disease.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

WB: The detection limit for TLR5 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: TLR7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat