Specifications
- Antibody type:Primary
- Antigen name:Serum/glucocorticoid regulated kinase 1
- Antigen symbol:SGK1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Gene ID:6446
- Antigen synonyms:SGK
- UniProtKB:O00141
- Storage temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human SGK1 (28-55aa MKQRRMGLNDFIQKIANNSYACKHPEVQ), different from the related mouse sequence by three amino acids, and from the related rat sequence by one amino acid.
- Purification:Immunogen affinity purified.
- Size:100 µg/vial
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Serine/threonine-protein kinase Sgk1(SGK1) detection. Tested with WB in Human.
Serine/threonine-protein kinase Sgk1, also called SGK1, encodes a serine/threonine protein kinase that plays an important role in cellular stress response. By fluorescence in situ hybridization, this gene is mapped to 6q23.2. This kinase activates certain potassium, sodium, and chloride channels, suggesting an involvement in the regulation of processes such as cell survival, neuronal excitability, and renal sodium excretion. High levels of expression of this gene may contribute to conditions such as hypertension and diabetic nephropathy. Several alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
WB: The detection limit for SGK1 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: SLC2A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat