Order Entry
Export
ContactUsLinkComponent
Anti-STIM1 Rabbit Polyclonal Antibody
Anti-STIM1 Rabbit Polyclonal Antibody
Catalog # BSBTPB9406
Supplier:  Boster Bio
undefined
Anti-STIM1 Rabbit Polyclonal Antibody
Catalog # BSBTPB9406
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Stromal interaction molecule 1
  • Antigen symbol:
    STIM1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Gene ID:
    6786
  • Antigen synonyms:
    STRMK|IMD10|TAM|GOK|D11S4896E|TAM1
  • UniProtKB:
    Q13586
  • Storage temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human STIM1(45-74aa AAEFCRIDKPLCHSEDEKLSFEAVRNIHKL), identical to the related mouse and rat sequences.
  • Purification:
    Immunogen affinity purified.
  • Size:
    100 µg/vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Stromal interaction molecule 1(STIM1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Stromal interaction molecule 1 is a protein that in humans is encoded by the STIM1 gene. STIM1 has a single transmembranedomain, and is localized to the endoplasmic reticulum, and to a lesser extent to the plasma membrane. This gene encodes a type 1 transmembrane protein that mediates Ca2+ influx after depletion of intracellular Ca2+ stores by gating of store-operated Ca2+ influx channels (SOCs). It is one of several genes located in the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocrotical carcinoma, and lung, ovarian, and breast cancer. This gene may play a role in malignancies and disease that involve this region, as well as early hematopoiesis, by mediating attachment to stromal cells. Mutations in this gene are associated with fatal classic Kaposi sarcoma, immunodeficiency due to defects in store-operated calcium entry (SOCE) in fibroblasts, ectodermal dysplasia and tubular aggregate myopathy. This gene is oriented in a head-to-tail configuration with the ribonucleotide reductase 1 gene (RRM1), with the 3' end of this gene situated 1.6 kb from the 5' end of the RRM1 gene. Alternative splicing of this gene results in multiple transcript variants.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: STRAP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat