Specifications
- Antibody type:Primary
- Antigen name:Nucleophosmin
- Antigen symbol:NPM
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Gene ID:4869
- Antigen synonyms:NPM|B23
- UniProtKB:P06748
- Storage temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Immunogen:E.coli-derived human Nucleophosmin recombinant protein (Position: M1-L294). Human Nucleophosmin shares 95% amino acid (aa) sequence identity with both mouse and rat Nucleophosmin.
- Purification:Immunogen affinity purified.
- Size:100 µg/vial
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Nucleophosmin(NPM1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
NPM1(Nucleophosmin/Nucleoplasmin family, member1), also known as NPM, nucleolar phosphoprotein B23 or numatrin, is a protein that in humans is encoded by the NPM1 gene. The NPM1 gene maps to chromosome 5q35. Chan et al. (1989) found that nucleophosmin is a nucleolar phosphoprotein that is more abundant in tumor cells than in normal resting cells. Stimulation of the growth of normal cells, e.g., mitogen activation of B lymphocytes, was accompanied by an increase in nucleophosmin protein level. They stated that nucleophosmin is likely involved in the assembly of ribosomal proteins into ribosomes. Electron microscopic study indicated that nucleophosmin is concentrated in the granular region of the nucleolus, where ribosome assembly occurs.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
WB: The detection limit for Nucleophosmin is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: NR3C1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat