Anti-Connexin 37/GJA4 Rabbit Antibody
Catalog # BSBTPA2205
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Connexin 37
- Antigen symbol:Cx37
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:near the N-terminus
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:Cx37|Gja4|Gap junction alpha-4 protein|Cxn-37
- Amino acid number:3 - 17
- Sequence:DWGFLEKLLDQVQEH
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human Connexin 37 (3-17aa DWGFLEKLLDQVQEH), identical to the related mouse and rat sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Gap junction alpha-4 protein(GJA4) detection. Tested with WB, IHC-P, ICC in Human, Mouse, Rat.
Reconstitute with distilled water
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Connexin 37/GJA4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat