Order Entry
Export
ContactUsLinkComponent
Anti-CSEN Rabbit Antibody
Anti-CSEN Rabbit Antibody
  BSBTPA2272
undefined
Anti-CSEN Rabbit Antibody
  BSBTPA2272

 

  • Antibody type:
    Primary
  • Antigen symbol:
    CSEN
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Amino acid number:
    1 - 16
  • Sequence:
    MQPAKEVTKASDGSLL
  • Storage temperature:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human CSEN (1-16aa MQPAKEVTKASDGSLL).
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Calsenilin(KCNIP3) detection. Tested with WB in Human.

Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CSEN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: