Anti-TMEM16A Rabbit Antibody
Catalog # BSBTPA2290
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Anoctamin 1
- Antigen symbol:ANO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:ORAOV2|DOG1|Transmembrane Protein 16A|TAOS2|TMEM16A
- Amino acid number:927 - 943
- Sequence:KVLMVELFMREEQDKQQ
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human TMEM16A (927-943aa KVLMVELFMREEQDKQQ), different from the related mouse and rat sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Anoctamin-1(ANO1) detection. Tested with WB in Human, Mouse, Rat.
Reconstitute with distilled water
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: ANO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat