Industrial gloves, Gobi® 112
Supplier: KCL by Honeywell
Certificates
About this item
This high quality cotton jersey liner with special nitrile coating meets all the requirements of a fully-coated protective glove with a wide usage spectrum: In addition to mechanical protection it also offers protection against liquids, high touch sensitivity and high temperature resistance.
- Wide cuff for quick donning and doffing
- Excellent mechanical resistance, moisture-repellent
- Good grip when handling wet parts
- Skin-friendly, high-quality cotton liner
- Generous length for additional forearm protection
Ideal for metalworking and mechanical engineering, packaging and transportation works, building and construction, when working with oily and wet parts and for the plastics industry.
Certifications: EN 388 (3111), Cat. II
Specifications
- Catalog No:
- KCHL112.9
- KCHL112.10
- Model:
- 112
- 112
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Finish:
- Fully coated
- Fully coated
- Cuff style:
- Gauntlet
- Gauntlet
- EN norm:
- EN 388 (3111)
- EN 388 (3111)