Order Entry
Export
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # COBSD5-112-FC
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # COBSD5-112-FC

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    RPE (R-Phycoerythrin)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Cross adsorption:
    Bovine, Rabbit
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse